Learn More
Cayman Chemical MMAI (hydrochloride), Purity: ≥98%, CAS: 132980-17-7, 50 mg
SDP
Catalog No. 502954265
Encompass_Preferred
A psychoactive aminoindan which can be found in recreational drug mixtures; potently inhibits serotonin uptake (IC50 = 212 nM) and stimulates its release without inducing neurotoxicity
Catalog No. 50-295-4265 Supplier Cayman Chemical Supplier No. 1389550 MG
A psychoactive aminoindan which can be found in recreational drug mixtures; potently inhibits serotonin uptake (IC50 = 212 nM) and stimulates its release without inducing neurotoxicity