Learn More
Andwin Scientific MAILING TUBE 5ML 500/PK
SDP
Catalog No. NC9319302
Encompass_Preferred
With leakproof feature, the Sample Mailing Tubes are ideal for shipping. This product is autoclavable, disposable, and economical. There are graduations in 1 mL increments. There is a frosted writing surface that permits users to label and organize t
Catalog No. NC9319302 Supplier Andwin Scientific Supplier No. 16465262PK
With leakproof feature, the Sample Mailing Tubes are ideal for shipping. This product is autoclavable, disposable, and economical. There are graduations in 1 mL increments. There is a frosted writing surface that permits users to label and organize t