Learn More
MSC Partners Brand Mailing Tubes, 3 x 25 in, Kraft, 24/Pack
Catalog No. 501915534
Encompass
These mailing tubes are designed for securely packaging and shipping various items. Made from durable spiral wound fibreboard, they offer reliable protection during transit and come with caps for complete enclosure.
  • Made from 0.07 thick spiral wound fibreboard
  • Kraft color
  • Includes caps for secure closure
  • Package quantity of 24
Catalog No. 50-191-5534 Supplier MSC Supplier No. 39550330
These mailing tubes are designed for securely packaging and shipping various items. Made from durable spiral wound fibreboard, they offer reliable protection during transit and come with caps for complete enclosure.
  • Made from 0.07 thick spiral wound fibreboard
  • Kraft color
  • Includes caps for secure closure
  • Package quantity of 24