Learn More
Chemglass Life Sciences Main Trap Body Only (D), Airfree, Schlenk
SDP
Catalog No. AF020305
Encompass_Preferred
Main Trap Body Only (D), Airfree, Schlenk. Dual trap assembly for use with AF-0060 or AF-0061 manifolds. The main trap (D) is constructed using a 57mm flat o-ring flange and the body is 57mm OD x 250mm long. Center to center distance between the traps is 175mm. Trap assembly is designed to allow connection to either the right or left side of AF-0060 or AF-0061 manifolds. Complete item is supplied with one #331 Viton o-ring, one CG-139-02 horseshoe clamp, three #116 Viton o-rings and two CG-150-05 #28 Stainless Steel pinch clamps.
Catalog No. AF020305 Supplier Chemglass Life Sciences Supplier No. AF020305
Main Trap Body Only (D), Airfree, Schlenk. Dual trap assembly for use with AF-0060 or AF-0061 manifolds. The main trap (D) is constructed using a 57mm flat o-ring flange and the body is 57mm OD x 250mm long. Center to center distance between the traps is 175mm. Trap assembly is designed to allow connection to either the right or left side of AF-0060 or AF-0061 manifolds. Complete item is supplied with one #331 Viton o-ring, one CG-139-02 horseshoe clamp, three #116 Viton o-rings and two CG-150-05 #28 Stainless Steel pinch clamps.