Learn More
Chemglass Life Sciences Main Trap Stopper Assembly (C), Airfree, Schlenk
SDP
Catalog No. 501530946
Encompass_Preferred
Main Trap Stopper Assembly (C), Airfree, Schlenk. Dual trap assembly for use with AF-0060 or AF-0061 manifolds. Pre-trap (B) is 51mm OD x 250mm long with 28/15 polished socket joints to connect the line shut off adapter (A) to the main trap stopper assembly (C) which has 28/15 o-ring ball joints. Trap assembly is designed to allow connection to either the right or left side of AF-0060 or AF-0061 manifolds.
Catalog No. 50-153-0946 Supplier Chemglass Life Sciences Supplier No. AF020304
Main Trap Stopper Assembly (C), Airfree, Schlenk. Dual trap assembly for use with AF-0060 or AF-0061 manifolds. Pre-trap (B) is 51mm OD x 250mm long with 28/15 polished socket joints to connect the line shut off adapter (A) to the main trap stopper assembly (C) which has 28/15 o-ring ball joints. Trap assembly is designed to allow connection to either the right or left side of AF-0060 or AF-0061 manifolds.