Learn More
Thermo Scientific Seal Kit for Pump 2 Pieces
Supplier:  Thermo Scientific 6040.0304
Encompass
PTFE piston seal kit with two replacement seals compatible with Thermo Scientific™ HPLC pumps designed to provide a leak-free seal between pump head and piston engineered to precisely fit SD pump model replacing 6040.0304 part number.
Catalog No. 50-134-7775