Learn More
Maine Molecular Quality Controls Inc BioFire RP2.1/RP2.1plus Control Panel M441
SDP
Catalog No. NC1812062
Encompass
BioFire RP2.1/RP2.1plus Control Panel M441 is intended for use as an external positive and negative assayed quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection on the BioFire Respiratory Panel 2.1 (RP2.1), BioFire Respiratory Panel 2.1plus (RP2.1plus) and BioFire Respiratory Panel 2.1-EZ (RP2.1-EZ) assays performed on the BioFire FilmArray systems. BioFire RP2.1/RP2.1plus Positive control is composed of synthetic RNA transcripts specifically designed for and intended to be used solely with the BioFire RP2.1, BioFire RP2.1plus and BioFire RP2.1-EZ assays. The BioFire RP2.1/RP2.1plus Control Panel M441 is comprised of 12 single use tubes, 6 tubes of BioFire RP2.1/RP2.1plus Positive and 6 tubes of BioFire RP2.1/RP2.1plus Negative, 300µL each.
Catalog No. NC1812062 Supplier Maine Molecular Quality Controls Inc Supplier No. M441
BioFire RP2.1/RP2.1plus Control Panel M441 is intended for use as an external positive and negative assayed quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection on the BioFire Respiratory Panel 2.1 (RP2.1), BioFire Respiratory Panel 2.1plus (RP2.1plus) and BioFire Respiratory Panel 2.1-EZ (RP2.1-EZ) assays performed on the BioFire FilmArray systems. BioFire RP2.1/RP2.1plus Positive control is composed of synthetic RNA transcripts specifically designed for and intended to be used solely with the BioFire RP2.1, BioFire RP2.1plus and BioFire RP2.1-EZ assays. The BioFire RP2.1/RP2.1plus Control Panel M441 is comprised of 12 single use tubes, 6 tubes of BioFire RP2.1/RP2.1plus Positive and 6 tubes of BioFire RP2.1/RP2.1plus Negative, 300µL each.