Learn More
Parker Hannifan Parker MK7694 Annual Maintenance Kit, Ensures Optimal Generator Performance, Extends Generator Lifespan, Unit: 1 Kit
SDP
Catalog No. NC9178352
Encompass
Parker Hannifin MK7694 Annual Maintenance Kit ensures optimal performance for Parker Balston UHP Nitrogen Generators. This essential kit supports the routine replacement of key filtration components to maintain nitrogen purity and operational efficiency while extending system lifespan. Regular maintenance helps reduce contamination risks, distinguishing it from other products by providing a comprehensive solution tailored for specific generator models.
Catalog No. NC9178352 Supplier Parker Hannifan Supplier No. MK7694
Parker Hannifin MK7694 Annual Maintenance Kit ensures optimal performance for Parker Balston UHP Nitrogen Generators. This essential kit supports the routine replacement of key filtration components to maintain nitrogen purity and operational efficiency while extending system lifespan. Regular maintenance helps reduce contamination risks, distinguishing it from other products by providing a comprehensive solution tailored for specific generator models.