Learn More
Zen Bio Inc MAINTAINANCE MEDIA 500ML
SDP
Catalog No. NC9860642
Encompass
Cell Culture Media; Adipocyte Medium; Zen-Bio; Maintenance medium after differentiation; Contains all factors necessary to support lipid accumulation in the maturing adipocytes; Ready-to-use; 500mL
Catalog No. NC9860642 Supplier Zen Bio Inc Supplier No. AM1
Cell Culture Media; Adipocyte Medium; Zen-Bio; Maintenance medium after differentiation; Contains all factors necessary to support lipid accumulation in the maturing adipocytes; Ready-to-use; 500mL