Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Maintaining the Human Body Flip Chart with Online Multimedia Lesson
SCI_ED

Catalog No. S81115 Shop All NewPath Learning Products
Change view
Click to view available options
Format:
Download
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S81115 Download
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81115 Supplier NewPath Learning™ Supplier No. 346753

missing translation for 'validMainframeMsg'

Combine illuminating graphics and write-on/wipe-off exercises with multimedia lessons to master key concepts on the human body.

  • Provides graphical instruction and write-on/wipe-off activities with one-year online access to a corresponding Multimedia Lesson featuring in-depth, interactive coverage of the concept
  • The Multimedia Lesson can be accessed by a teacher and ALL of your students anywhere, anytime, on any device – including Chromebooks, tablets, computers and mobile devices.
  • Flip Chart Set features 10 Laminated, Double-Sided Charts
  • Includes a Teacher Guide with copymaster activities and review questions
  • Each lesson features Animated, Narrated Tutorials; Interactive Exercises & Activities; Virtual Lab Investigations; Illustrated, Narrated Glossary; Visual Resource Library; and an Inquiry-Based Student Activity Guide
  • Developed under sponsorship of the National Institutes of Health (NIH), the lessons are perfect for individual student instruction on any computer as well as classroom presentation on any Interactive Whiteboard
  • Covers: Disease & the Body's Defenses; Inflammation; Immune Response; Illness; Immunity/Allergies; Skin; Male/Female Reproductive Systems; Fertilization & Fetal Development; and Systems Working Together

Specifications

Grade 6 to 10
Description NGSS Flip Chart with Online Lesson
Category Biology
Format Download
For Use With (Application) Classroom Instruction
Includes Flip Chart, Online Lesson
License Site
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.