Learn More
Hach Company Maintenance Kit for 5500sc Silica Analyzer, 2/4 Channel - Hach (MPN 6788302)
Catalog No. NC3357119 Shop All Hach Company Products
Encompass
This maintenance kit provides the replacement parts and consumables required for scheduled maintenance of 5500sc silica analyzers, supporting 2- and 4-channel configurations to help maintain fluid handling and measurement accuracy.
  • Includes replacement consumables and wear parts for routine service.
  • Contains fluid handling components to preserve measurement integrity.
  • Provides colorimeter cell and stir bar for optical measurement maintenance.
  • Includes filters, fittings, and check valves to support reliable operation.
  • Supplies labels and instructions to simplify scheduled maintenance tasks.
Catalog No. NC3357119 Supplier Hach Company Supplier No. 6788302
This maintenance kit provides the replacement parts and consumables required for scheduled maintenance of 5500sc silica analyzers, supporting 2- and 4-channel configurations to help maintain fluid handling and measurement accuracy.
  • Includes replacement consumables and wear parts for routine service.
  • Contains fluid handling components to preserve measurement integrity.
  • Provides colorimeter cell and stir bar for optical measurement maintenance.
  • Includes filters, fittings, and check valves to support reliable operation.
  • Supplies labels and instructions to simplify scheduled maintenance tasks.