Learn More
Parker Hannifan Parker Hannifin MK7572C Maintenance Kit, Ensures High Nitrogen Purity 95-99.5%, Quick Service Time 15 Minutes, Unit: Kit
SDP
Catalog No. NC9797145
Encompass
The Parker Hannifin MK7572C is a 12-month maintenance kit designed for optimal performance of Parker Nitrogen Gas Generators, ensuring high purity nitrogen output (95% to 99.5%). This kit includes essential filtration components that effectively remove contaminants, prolonging the lifespan of the generators and maintaining quality output. With a service time of just 15 minutes, it differentiates itself through its efficiency and specific compatibility with multiple nitrogen generator models.
Catalog No. NC9797145 Supplier Parker Hannifan Supplier No. MK7572C
The Parker Hannifin MK7572C is a 12-month maintenance kit designed for optimal performance of Parker Nitrogen Gas Generators, ensuring high purity nitrogen output (95% to 99.5%). This kit includes essential filtration components that effectively remove contaminants, prolonging the lifespan of the generators and maintaining quality output. With a service time of just 15 minutes, it differentiates itself through its efficiency and specific compatibility with multiple nitrogen generator models.