Learn More
Hach Company Maintenance kit, high range phosphate, 2/4 channels - for Hach 5500sc and similar high-range phosphate analyzers
Catalog No. NC3361134 Shop All Hach Company Products
Encompass
This maintenance kit provides replacement consumables and parts for routine servicing of high-range phosphate analyzers in 2- or 4-channel configurations. It helps maintain measurement accuracy and instrument uptime.
  • Includes air filter, tubing, and color-coded bottle caps.
  • Includes a colorimeter cell and stir bar for optical measurement maintenance.
  • Includes fittings, check valves, and a capillary assembly to replace worn fluid-path components.
  • Includes a pressure transducer and fan filter for stable operation.
  • Includes user instructions to guide installation and service procedures.
Catalog No. NC3361134 Supplier Hach Company Supplier No. 6788310
This maintenance kit provides replacement consumables and parts for routine servicing of high-range phosphate analyzers in 2- or 4-channel configurations. It helps maintain measurement accuracy and instrument uptime.
  • Includes air filter, tubing, and color-coded bottle caps.
  • Includes a colorimeter cell and stir bar for optical measurement maintenance.
  • Includes fittings, check valves, and a capillary assembly to replace worn fluid-path components.
  • Includes a pressure transducer and fan filter for stable operation.
  • Includes user instructions to guide installation and service procedures.