Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Tip Seal Replacement Kit, HiScroll 6, OEM Maintenance
Catalog No. NC3978975
Encompass_Preferred
This OEM maintenance kit is for the Pfeiffer Vacuum HiScroll 6, an oil-free, dry scroll pump with IPM synchronous motors for increased efficiency and lower operating temperatures. Designed for compact systems, it features adaptive cooling and noise reduction.
  • OEM maintenance kit for dry scroll pumps.
  • IPM synchronous motors offer up to 15% greater efficiency.
  • Ensures lower operating temperatures with reduced heat generation.
  • Ideal for compact systems needing high cooling efficiency.
  • Incorporates adaptive cooling control.
  • Reduces noise by throttling pump speed when full performance is not required.
  • Supports intelligent speed control and monitoring via optional integrated pressure sensor.
  • Automatically regulates pumping speed to hold specific pressures, extending maintenance intervals.
  • Compatible with other products via RS-485 or ProfiNet interface.
Catalog No. NC3978975 Supplier PFEIFFER VACUUM INC Supplier No. 2000026535
This OEM maintenance kit is for the Pfeiffer Vacuum HiScroll 6, an oil-free, dry scroll pump with IPM synchronous motors for increased efficiency and lower operating temperatures. Designed for compact systems, it features adaptive cooling and noise reduction.
  • OEM maintenance kit for dry scroll pumps.
  • IPM synchronous motors offer up to 15% greater efficiency.
  • Ensures lower operating temperatures with reduced heat generation.
  • Ideal for compact systems needing high cooling efficiency.
  • Incorporates adaptive cooling control.
  • Reduces noise by throttling pump speed when full performance is not required.
  • Supports intelligent speed control and monitoring via optional integrated pressure sensor.
  • Automatically regulates pumping speed to hold specific pressures, extending maintenance intervals.
  • Compatible with other products via RS-485 or ProfiNet interface.