Learn More
Hach Company Hach Maintenance Kit, Low Range Phosphate (2/4 Channels) - Replacement consumables for low-range phosphate analyzers
Catalog No. NC3357120 Shop All Hach Company Products
Encompass
This maintenance kit supplies the routine replacement consumables and wearable components used during preventive service of low-range phosphate analyzers configured for two or four channels. It restores measurement performance and helps reduce downtime for continuous orthophosphate monitoring.
  • Includes common replacement parts and consumables for preventive maintenance.
  • Contents include air filter, tubing, color-coded bottle caps, colorimeter cell, stir bar, fittings, pressure transducer, check valves, and capillary assembly.
  • Also contains fan filter and user instructions for straightforward replacement.
  • Designed for 2- or 4-channel low-range phosphate analyzer configurations.
  • Helps restore analyzer reliability and maintain accurate low-range phosphate measurements.
Catalog No. NC3357120 Supplier Hach Company Supplier No. 6788308
This maintenance kit supplies the routine replacement consumables and wearable components used during preventive service of low-range phosphate analyzers configured for two or four channels. It restores measurement performance and helps reduce downtime for continuous orthophosphate monitoring.
  • Includes common replacement parts and consumables for preventive maintenance.
  • Contents include air filter, tubing, color-coded bottle caps, colorimeter cell, stir bar, fittings, pressure transducer, check valves, and capillary assembly.
  • Also contains fan filter and user instructions for straightforward replacement.
  • Designed for 2- or 4-channel low-range phosphate analyzer configurations.
  • Helps restore analyzer reliability and maintain accurate low-range phosphate measurements.