Learn More
Medchemexpress LLC MAIT-203 (acetate) | 99.1% | 2577.90 | 25 MG
SDP
Catalog No. 5000425484
Encompass_Preferred
MAIT-203 acetate is a cyclopentyalanin-derived peptidomimetic that potently inhibits the interaction of adenomatous polyposis coli (APC) and Asef (RhoGEF4), but not APC-Sam68 or APC-striatin interactions. It binds APC-ARM with a Ki value of 0.015 μM and a Kd value of 0.036 μΜ. This compound significantly represses the migration and invasion of colorectal cancer cells.
  • Potently inhibits APC-Asef interaction
  • Binds APC-ARM with a Ki value of 0.015 μM and a Kd value of 0.036 μΜ
  • Significantly represses migration and invasion of colorectal cancer cells
  • Reduces Asef-APC interaction in HEK293T cells
  • Attenuates SW480 and HCT116 cell migration
  • Decreases the active form of CDC42
Catalog No. 50-004-25484 Supplier Medchemexpress LLC Supplier No. 25MGHYP2269A
MAIT-203 acetate is a cyclopentyalanin-derived peptidomimetic that potently inhibits the interaction of adenomatous polyposis coli (APC) and Asef (RhoGEF4), but not APC-Sam68 or APC-striatin interactions. It binds APC-ARM with a Ki value of 0.015 μM and a Kd value of 0.036 μΜ. This compound significantly represses the migration and invasion of colorectal cancer cells.
  • Potently inhibits APC-Asef interaction
  • Binds APC-ARM with a Ki value of 0.015 μM and a Kd value of 0.036 μΜ
  • Significantly represses migration and invasion of colorectal cancer cells
  • Reduces Asef-APC interaction in HEK293T cells
  • Attenuates SW480 and HCT116 cell migration
  • Decreases the active form of CDC42