Learn More
MSC Majestic Glove Korplex Sleeve, 14 in, ANSI A4, 1 ea
Catalog No. 5000788939
Encompass
Majestic Glove Cut-Less Korplex Cut Resistant Sleeves are engineered with Korplex, a patented yarn that is cool to the touch and provides exceptional cut resistance. These sleeves are 14 in. long, feature a convenient thumbhole, and offer an ANSI A4 cut resistance level in a high-visibility yellow.
  • Korplex® cut resistant technology
  • Seamless knit construction for comfortable fit
  • Length: 14 in.
  • High-visibility yellow
  • Thumbhole cuff
  • ANSI A4 cut resistance
  • EN 388: 4542 certified
Catalog No. 50-007-88939 Supplier MSC Supplier No. 20228136
Majestic Glove Cut-Less Korplex Cut Resistant Sleeves are engineered with Korplex, a patented yarn that is cool to the touch and provides exceptional cut resistance. These sleeves are 14 in. long, feature a convenient thumbhole, and offer an ANSI A4 cut resistance level in a high-visibility yellow.
  • Korplex® cut resistant technology
  • Seamless knit construction for comfortable fit
  • Length: 14 in.
  • High-visibility yellow
  • Thumbhole cuff
  • ANSI A4 cut resistance
  • EN 388: 4542 certified