Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit - Alcatel-Adixen 2005
Catalog No. 501956222
Encompass_Preferred
Major Repair Kits include o-rings, gaskets, washers, shaft seals, vanes, springs sight glass, and coupling element for major overhaul
Catalog No. 50-195-6222 Supplier Precision Plus Vacuum Parts Inc Supplier No. 65885FR
Major Repair Kits include o-rings, gaskets, washers, shaft seals, vanes, springs sight glass, and coupling element for major overhaul