Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit - Alcatel-Adixen 2010C1
Catalog No. 501956201
Encompass_Preferred
Major Repair Kits include o-rings, gaskets, washers, shaft seals, vanes, springs sight glass, and coupling element for major overhaul
Catalog No. 50-195-6201 Supplier Precision Plus Vacuum Parts Inc Supplier No. 104977FR
Major Repair Kits include o-rings, gaskets, washers, shaft seals, vanes, springs sight glass, and coupling element for major overhaul