Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit - Alcatel-Adixen 2020A
Catalog No. 501956212
Encompass_Preferred
Major Repair Kits include o-rings, gaskets, washers, shaft seals, vanes, springs sight glass, and coupling element for major overhaul
Catalog No. 50-195-6212 Supplier Precision Plus Vacuum Parts Inc Supplier No. 52982FR
Major Repair Kits include o-rings, gaskets, washers, shaft seals, vanes, springs sight glass, and coupling element for major overhaul