Learn More
Precision Plus Vacuum Parts Inc Alcatel Major Kit - AP122L
Catalog No. 501956180
Encompass_Preferred
Alcatel Major Kit for models AP122L includes o-rings, bearings, grease, and wave washers for major overhaul
Catalog No. 50-195-6180 Supplier Precision Plus Vacuum Parts Inc Supplier No. 110910
Alcatel Major Kit for models AP122L includes o-rings, bearings, grease, and wave washers for major overhaul