Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit for Leybold D4A
Catalog No. 501956183
Encompass_Preferred
Major Repair Kit includes coupling element, o-rings, gaskets, shaft seals, sight glass, and vanes for major pump maintenance
Catalog No. 50-195-6183 Supplier Precision Plus Vacuum Parts Inc Supplier No. 10167KIT
Major Repair Kit includes coupling element, o-rings, gaskets, shaft seals, sight glass, and vanes for major pump maintenance