Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit for Leybold D8A
Catalog No. 501956184
Encompass_Preferred
Major Repair Kit includes coupling element, o-rings, gaskets, shaft seals, sight glass, and vanes for major pump maintenance
Catalog No. 50-195-6184 Supplier Precision Plus Vacuum Parts Inc Supplier No. 10168KIT
Major Repair Kit includes coupling element, o-rings, gaskets, shaft seals, sight glass, and vanes for major pump maintenance