Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit for Pfeiffer UNO35
Catalog No. 501956238
Encompass_Preferred
Major Repair Kit includes o-rings, shaft seals, leak nozzle, hydraulic pistons, sight glass, sight glass gasket, springs, valves, and vanes for major pump maintenance
Catalog No. 50-195-6238 Supplier Precision Plus Vacuum Parts Inc Supplier No. PKE22001T
Major Repair Kit includes o-rings, shaft seals, leak nozzle, hydraulic pistons, sight glass, sight glass gasket, springs, valves, and vanes for major pump maintenance