Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit for Scroll Pump - Ulvac DIS90, Anest-Iwata ISP90, Leybold SC5D
Catalog No. 501956255
Encompass_Preferred
Major Rebuild Kit includes tip seals, crank pins, bearings, o-rings, shaft seals, and other components for major pump maintenance
Catalog No. 50-195-6255 Supplier Precision Plus Vacuum Parts Inc Supplier No. 98882590
Major Rebuild Kit includes tip seals, crank pins, bearings, o-rings, shaft seals, and other components for major pump maintenance