Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit for Pfeiffer UNO1.5A and UNO2.5A
Catalog No. 501956229
Encompass_Preferred
Major Repair Kit for UNO 1.5A and UNO 2.5A includes sight glass, oil box gasket, shaft seal, valve plate, flap valve, centering ring, vane springs, locking caps, and o-rings * Does not contain vanes
Catalog No. 50-195-6229 Supplier Precision Plus Vacuum Parts Inc Supplier No. 080010
Major Repair Kit for UNO 1.5A and UNO 2.5A includes sight glass, oil box gasket, shaft seal, valve plate, flap valve, centering ring, vane springs, locking caps, and o-rings * Does not contain vanes