Learn More
Waters Corp Major Mix IMS/Tof Calibration Kit
Catalog No. 501384617 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Major Mix IMS/Tof Calibration Kit is intended to be used as a calibration solution for high resolution mass spectrometers equipped with ion mobility. This kit is also utilized in the on-board calibration of the Vion IMS QTof system.
Catalog No. 50-138-4617 Supplier Waters Corp Supplier No. 186008113
The Major Mix IMS/Tof Calibration Kit is intended to be used as a calibration solution for high resolution mass spectrometers equipped with ion mobility. This kit is also utilized in the on-board calibration of the Vion IMS QTof system.