Learn More
TARGETMOL CHEMICALS INC MAK-683 HYDROCHLORIDE 5MG
SDP
Catalog No. 502578975
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. MAK683 hydrochloride is an inhibitor of embryonic ectoderm development (EED) (patent US20160176882 A1 compound example 2). MAK683 exhibits IC50s of 59 26 89 nM in EED Alphascreen binding ELISA and LC-MS assay. purity: 98%
Catalog No. 50-257-8975 Supplier TARGETMOL CHEMICALS INC Supplier No. T96815MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. MAK683 hydrochloride is an inhibitor of embryonic ectoderm development (EED) (patent US20160176882 A1 compound example 2). MAK683 exhibits IC50s of 59 26 89 nM in EED Alphascreen binding ELISA and LC-MS assay. purity: 98%