Learn More
Aviva Systems Biology Corp MAK10 Antibody - N-terminal region (ARP46333_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46333_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAK10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAK10. Alternate names: FLJ21613, FLJ22643, bA379P1.1, MAK10, RP11-379P1.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59306