Learn More
Medchemexpress LLC HY-103663 10mg , MAK683 CAS:1951408-58-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10366310MG
Encompass_Preferred
HY-103663 10mg MAK683 CAS: 1951408-58-4 MAK683 is an embryonic ectoderm development (EED) inhibitor extracted from patent US20160176882 A1, compound example 2. MAK683 exhibits IC50s of 59, 89, 26 nM in EED Alphascreen binding, LC-MS and ELISA assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0449