Learn More
Description
- Foster creativity and innovation with engineering concepts using our teacher-curated collection of makerspace materials.
Specifications
Specifications
| Grade Level | Elementary |
| Grade | K to 5 |
| Class Size | 1 Class |
| Product Type | Makerspace Stem Lab Materials |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
