Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Makerspace STEM Lab Materials 3BX
SCI_ED

Catalog No. p-200022148
Change view
Click to view available options
Grade:
K to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02755 K to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02755 Supplier Hand2Mind Supplier No. 92930
Only null left
Add to Cart
Add to Cart

Foster creativity and innovation with engineering concepts using our teacher-curated collection of makerspace materials.

  • Foster creativity and innovation with engineering concepts using our teacher-curated collection of makerspace materials.

Specifications

Grade Level Elementary
Grade K to 5
Class Size 1 Class
Product Type Makerspace Stem Lab Materials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.