Learn More
Abcam Makisterone A, EcR agonist, 1MG
Catalog No. 502705913
Encompass_Preferred
MW 494.7 Da, Purity >95%. Potent ecdysone receptor (EcR) agonist. Shows similar affinity for EcR as 20-Hydroxyecdysone (ab142425) Predominant insect moulting hormone in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5913 Supplier Abcam Supplier No. AB1442951MG
MW 494.7 Da, Purity >95%. Potent ecdysone receptor (EcR) agonist. Shows similar affinity for EcR as 20-Hydroxyecdysone (ab142425) Predominant insect moulting hormone in vivo. .
The product is subject to the following: Abcam Restricted Use Statement