Learn More
Qiagen Beverly Inc MAKODNA POLYMERASE(3 -5 EXO-)
Catalog No. 50305915
Encompass
DNA Polymerase; Mako DNA Polymerase (3'-5' Exo-); Conc. 30,000 U/ul; 3,000 U; incl. 10X buffer
Catalog No. 50-305-915 Supplier Qiagen Beverly Inc Supplier No. P7090L
DNA Polymerase; Mako DNA Polymerase (3'-5' Exo-); Conc. 30,000 U/ul; 3,000 U; incl. 10X buffer