Learn More
Sigma Aldrich MakroPor, silicon membrane
Catalog No. 503124476 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 Ea.
Catalog No. Quantity
50-312-4476 5 Ea.
1 options
Catalog No. 50-312-4476 Supplier Sigma Aldrich Supplier No. MP2501055EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Macro porous silicon is useful for applications requiring mechanical and chemical stability with high order of the pores. Our macro porous silicon membranes are manufactured from single crystal n-doped silicon wafers and possess pore diameter between 1 to 8 mum. The standard deviation of pore diameter and interpore distance for these porous silicon membrane materials is less than 5 %. These macro porous silicon finds applications in filtration, as platforms for multi-functional flow-through sensors, as fuels cells electrodes or micro reactors etc. They can also be used as templates for the fabrication of micro scale composites, such as tubes or wires by electrochemical deposition or by using polymer melts.Product of SmartMembranes-GmbH
Specifications
Linear Formula Si
Quantity 5 Ea.