Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-pab-exatecan | 3059991-66-8 | 96.1% | 1852.91 | C87H118FN9O34 | 25 MG
SDP

Catalog No. 5000203645
Encompass_Preferred

Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-pab-exatecan is an exatecan-containing ADC linker-payload intermediate intended for research use. It is supplied as an off-white to light yellow solid and is characterized by a high molecular weight and analytical purity suitable for biochemical and ADC development studies.

  • Molecular weight: 1852.91
  • Chemical formula: C87H118FN9O34
  • Purity: 96.1% (HPLC)
  • Appearance: off-white to light yellow solid
  • Storage: store at -20°C; in solvent, -80°C for up to 6 months

Catalog No. 50-002-03645 Supplier Medchemexpress LLC Supplier No. HY17363425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-pab-exatecan is an exatecan-containing ADC linker-payload intermediate intended for research use. It is supplied as an off-white to light yellow solid and is characterized by a high molecular weight and analytical purity suitable for biochemical and ADC development studies.

  • Molecular weight: 1852.91
  • Chemical formula: C87H118FN9O34
  • Purity: 96.1% (HPLC)
  • Appearance: off-white to light yellow solid
  • Storage: store at -20°C; in solvent, -80°C for up to 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.