Learn More
Medchemexpress LLC Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-pab-exatecan | 3059991-66-8 | 96.1% | 1852.91 | C87H118FN9O34 | 50 MG
SDP
Catalog No. 5000203287
Encompass_Preferred
Maleimide-PEG12 β-Glu-PAB-Exatecan is a drug-linker intermediate designed for antibody-drug conjugate (ADC) synthesis. The molecule contains a maleimide functional group for thiol conjugation, a PEG12 spacer to increase hydrophilicity and spacing, and a β-glutamate PAB linkage to an exatecan payload. The material is high molecular weight and supplied as a research quantity.
  • Drug-linker intermediate for ADC synthesis.
  • Maleimide functional group for thiol conjugation.
  • PEG12 spacer provides hydrophilicity and spacing.
  • β-glu-PAB cleavable linker enables payload release.
  • High molecular weight and analytical purity suitable for research use.
Catalog No. 50-002-03287 Supplier Medchemexpress LLC Supplier No. HY17363450MG
Add to Cart
Add to Cart
Request a Quote
Maleimide-PEG12 β-Glu-PAB-Exatecan is a drug-linker intermediate designed for antibody-drug conjugate (ADC) synthesis. The molecule contains a maleimide functional group for thiol conjugation, a PEG12 spacer to increase hydrophilicity and spacing, and a β-glutamate PAB linkage to an exatecan payload. The material is high molecular weight and supplied as a research quantity.
  • Drug-linker intermediate for ADC synthesis.
  • Maleimide functional group for thiol conjugation.
  • PEG12 spacer provides hydrophilicity and spacing.
  • β-glu-PAB cleavable linker enables payload release.
  • High molecular weight and analytical purity suitable for research use.