Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-pab-exatecan | 3059991-66-8 | 96.1% | 1852.91 g/mol | C87H118FN9O34 | 10 MG
SDP

Catalog No. 5000203602
Encompass_Preferred

Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-pab-exatecan is a research-grade drug-linker conjugate based on Exatecan for use in antibody-drug conjugate synthesis and preclinical research. Analytical documentation and safety data are provided by the manufacturer.

  • Research-grade ADC drug-linker conjugate for conjugation to antibodies or targeting moieties.
  • Purity 96.1% by HPLC.
  • Molecular weight 1852.91 g/mol.
  • Chemical formula C87H118FN9O34.
  • CAS number 3059991-66-8.
  • Available in small research packs, including 10 mg.
  • Long-term storage at -20°C; in solvent store at -80°C for up to 6 months.
  • Analytical documentation available: data sheet, COA, HNMR, RP-HPLC, LCMS, and SDS.

Catalog No. 50-002-03602 Supplier Medchemexpress LLC Supplier No. HY17363410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-pab-exatecan is a research-grade drug-linker conjugate based on Exatecan for use in antibody-drug conjugate synthesis and preclinical research. Analytical documentation and safety data are provided by the manufacturer.

  • Research-grade ADC drug-linker conjugate for conjugation to antibodies or targeting moieties.
  • Purity 96.1% by HPLC.
  • Molecular weight 1852.91 g/mol.
  • Chemical formula C87H118FN9O34.
  • CAS number 3059991-66-8.
  • Available in small research packs, including 10 mg.
  • Long-term storage at -20°C; in solvent store at -80°C for up to 6 months.
  • Analytical documentation available: data sheet, COA, HNMR, RP-HPLC, LCMS, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.