Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal((3S,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diamine-PEG12)-β-glu-PAB-exatecan | 3059991-66-8 | 96.1% | 1852.91 g·mol⁻¹ | C87H118FN9O34 | 5 MG
SDP

Catalog No. 5000202306
Encompass_Preferred

Exatecan-based ADC drug-linker conjugate intended for research use, combining a Topoisomerase I inhibitor with a cleavable β-glucuronide-PAB linker and a maleimide PEG12 spacer for antibody conjugation.

  • 96.1% purity.
  • Available in small quantities, including 5 mg.
  • Molecular weight 1852.91 g·mol⁻¹.
  • CAS number 3059991-66-8.
  • Chemical formula C87H118FN9O34.
  • Off-white to light yellow solid; store at -20 °C, protect from light, under inert gas.
  • In solvent: store at -80 °C up to 6 months, -20 °C up to 1 month.

Catalog No. 50-002-02306 Supplier Medchemexpress LLC Supplier No. HY1736345MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Exatecan-based ADC drug-linker conjugate intended for research use, combining a Topoisomerase I inhibitor with a cleavable β-glucuronide-PAB linker and a maleimide PEG12 spacer for antibody conjugation.

  • 96.1% purity.
  • Available in small quantities, including 5 mg.
  • Molecular weight 1852.91 g·mol⁻¹.
  • CAS number 3059991-66-8.
  • Chemical formula C87H118FN9O34.
  • Off-white to light yellow solid; store at -20 °C, protect from light, under inert gas.
  • In solvent: store at -80 °C up to 6 months, -20 °C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.