Learn More
Medchemexpress LLC Mal-amide-PEG2-oxyamine | 2253965-09-0 | 96.8% | 315.32 g/mol | C13H21N3O6 | 5 MG
SDP
Catalog No. 5000262263
Encompass_Preferred
Mal-amide-PEG2-oxyamine is a PEG-based bifunctional linker for assembling PROTACs and other bioconjugates. It features a maleimide amide and an oxyamine separated by a PEG2 spacer, permitting orthogonal thiol and oxime ligations. Supplied as a high-purity research reagent in milligram quantities.
  • Contains maleimide and oxyamine functional groups for orthogonal conjugation.
  • PEG2 spacer improves solubility and provides flexible linkage.
  • Designed for PROTAC synthesis and targeted bioconjugation applications.
  • High purity (96.8%) supports reproducible synthetic chemistry.
  • Available in small milligram pack sizes suitable for research.
  • Store pure material at -20°C for long-term stability.
Catalog No. 50-002-62263 Supplier Medchemexpress LLC Supplier No. HY1335075MG
Mal-amide-PEG2-oxyamine is a PEG-based bifunctional linker for assembling PROTACs and other bioconjugates. It features a maleimide amide and an oxyamine separated by a PEG2 spacer, permitting orthogonal thiol and oxime ligations. Supplied as a high-purity research reagent in milligram quantities.
  • Contains maleimide and oxyamine functional groups for orthogonal conjugation.
  • PEG2 spacer improves solubility and provides flexible linkage.
  • Designed for PROTAC synthesis and targeted bioconjugation applications.
  • High purity (96.8%) supports reproducible synthetic chemistry.
  • Available in small milligram pack sizes suitable for research.
  • Store pure material at -20°C for long-term stability.