Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-(CH2COOH)2 | 207613-14-7 | 99.2% | 284.22 | C11H12N2O7 | 10 MG
SDP

Catalog No. 5000202189
Encompass_Preferred

Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used in the synthesis of hydrophilic antibody-drug conjugate (ADC) linkers. It is a small-molecule building block supplied for research use only.

  • Contains maleimidoethyl functionality for conjugation reactions.
  • Suitable for hydrophilic ADC linker synthesis.
  • High reported purity (99.15%).
  • Molecular formula C11H12N2O7 and molecular weight 284.22.
  • Available in small mg-scale quantities for research applications.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-02189 Supplier Medchemexpress LLC Supplier No. HY2364210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used in the synthesis of hydrophilic antibody-drug conjugate (ADC) linkers. It is a small-molecule building block supplied for research use only.

  • Contains maleimidoethyl functionality for conjugation reactions.
  • Suitable for hydrophilic ADC linker synthesis.
  • High reported purity (99.15%).
  • Molecular formula C11H12N2O7 and molecular weight 284.22.
  • Available in small mg-scale quantities for research applications.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.