Learn More
Medchemexpress LLC Mal-amido-(CH2COOH)2 | 207613-14-7 | 99.2% | 284.22 | C11H12N2O7 | 100 MG
SDP
Catalog No. 5000202755
Encompass_Preferred
Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used as a hydrophilic linker building block in antibody-drug conjugate (ADC) synthesis. It is supplied as a white to off-white solid with high purity and is intended for research use only. Typical applications include conjugation chemistry and linker assembly where maleimide reactivity and carboxylic acid functionality are required.
  • Maleimide functional group for thiol conjugation.
  • Hydrophilic spacer with two carboxylic acid termini.
  • High purity suitable for research applications.
  • Available in multiple package sizes including 100 mg.
  • Characterized by 1H NMR and LC-MS data.
  • Comes with datasheet, COA, and SDS documentation.
Catalog No. 50-002-02755 Supplier Medchemexpress LLC Supplier No. HY23642100MG
Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used as a hydrophilic linker building block in antibody-drug conjugate (ADC) synthesis. It is supplied as a white to off-white solid with high purity and is intended for research use only. Typical applications include conjugation chemistry and linker assembly where maleimide reactivity and carboxylic acid functionality are required.
  • Maleimide functional group for thiol conjugation.
  • Hydrophilic spacer with two carboxylic acid termini.
  • High purity suitable for research applications.
  • Available in multiple package sizes including 100 mg.
  • Characterized by 1H NMR and LC-MS data.
  • Comes with datasheet, COA, and SDS documentation.