Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-(CH2COOH)2 | 207613-14-7 | 99.2% | 284.22 g·mol-1 | C11H12N2O7 | 50 MG
SDP

Catalog No. 5000202316
Encompass_Preferred

Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used in the synthesis of hydrophilic antibody-drug conjugate (ADC) linkers for research applications. It is supplied as a solid with high purity and is formulated to dissolve in DMSO for downstream conjugation workflows.

  • CAS number 207613-14-7.
  • Purity 99.2%.
  • Molecular formula C11H12N2O7.
  • Molecular weight 284.22 g·mol-1.
  • Appearance solid; soluble in DMSO with ultrasonic warming and heating to 60°C.
  • Storage powder -20°C up to 3 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Intended for research use only; not for human use.

Catalog No. 50-002-02316 Supplier Medchemexpress LLC Supplier No. HY2364250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used in the synthesis of hydrophilic antibody-drug conjugate (ADC) linkers for research applications. It is supplied as a solid with high purity and is formulated to dissolve in DMSO for downstream conjugation workflows.

  • CAS number 207613-14-7.
  • Purity 99.2%.
  • Molecular formula C11H12N2O7.
  • Molecular weight 284.22 g·mol-1.
  • Appearance solid; soluble in DMSO with ultrasonic warming and heating to 60°C.
  • Storage powder -20°C up to 3 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.