Learn More
Medchemexpress LLC Mal-amido-(CH2COOH)2 | 207613-14-7 | 99.2% | 284.22 g/mol | C11H12N2O7 | 5 MG
SDP
Catalog No. 5000203608
Encompass_Preferred
Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used in the synthesis of hydrophilic antibody-drug conjugate (ADC) linkers. It is provided as a research-use building block for linker chemistry.
  • Maleimidoethyl intermediate for ADC linker synthesis.
  • High reported purity supports consistent chemical performance.
  • Moderate molecular weight and polar functionality aid solubility in common organic solvents.
  • Reactive maleimide moiety enables conjugation to thiol-containing biomolecules.
  • Available in small research-scale quantities suitable for route development and linker optimization.
Catalog No. 50-002-03608 Supplier Medchemexpress LLC Supplier No. HY236425MG
Mal-amido-(CH2COOH)2 is a maleimidoethyl-containing intermediate used in the synthesis of hydrophilic antibody-drug conjugate (ADC) linkers. It is provided as a research-use building block for linker chemistry.
  • Maleimidoethyl intermediate for ADC linker synthesis.
  • High reported purity supports consistent chemical performance.
  • Moderate molecular weight and polar functionality aid solubility in common organic solvents.
  • Reactive maleimide moiety enables conjugation to thiol-containing biomolecules.
  • Available in small research-scale quantities suitable for route development and linker optimization.