Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-PEG1-C2-NHS ester | 1260092-50-9 | 99.7% | 381.34 | C16H19N3O8 | 50 MG
SDP

Catalog No. 5000201888
Encompass_Preferred

Mal-amido-PEG1-C2-NHS ester is a non-cleavable antibody-drug conjugate (ADC) linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester, used for bioconjugation to label primary amines on proteins, oligonucleotides, and other amine-containing molecules. The product is supplied as a high-purity, white to off-white solid and requires cold, inert storage to maintain stability.

  • Contains maleimide and NHS ester functional groups.
  • Enables selective labeling of primary amines.
  • High purity (≈99.7%).
  • Molecular weight 381.34 and formula C16H19N3O8.
  • Available in multiple sizes including 50 mg.
  • Store at -20°C, protect from light, under nitrogen; in solution: -80°C up to 6 months.

Catalog No. 50-002-01888 Supplier Medchemexpress LLC Supplier No. HY12650750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-amido-PEG1-C2-NHS ester is a non-cleavable antibody-drug conjugate (ADC) linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester, used for bioconjugation to label primary amines on proteins, oligonucleotides, and other amine-containing molecules. The product is supplied as a high-purity, white to off-white solid and requires cold, inert storage to maintain stability.

  • Contains maleimide and NHS ester functional groups.
  • Enables selective labeling of primary amines.
  • High purity (≈99.7%).
  • Molecular weight 381.34 and formula C16H19N3O8.
  • Available in multiple sizes including 50 mg.
  • Store at -20°C, protect from light, under nitrogen; in solution: -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.