Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-PEG1-C2-NHS ester | 1260092-50-9 | 99.7% | 381.34 g/mol | C16H19N3O8 | 10 MG
SDP

Catalog No. 5000203466
Encompass_Preferred

Mal-amido-PEG1-C2-NHS ester is a noncleavable ADC linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester, enabling conjugation to thiols and primary amines for antibody-drug conjugate construction and bioconjugation.

  • Contains maleimide and NHS ester functional groups for thiol and amine coupling.
  • Molecular weight approximately 381.34 g/mol.
  • Chemical formula C16H19N3O8; white to off-white solid.
  • High purity (listed as 99.69%) suitable for research applications.
  • Soluble in DMSO (100 mg/mL) with recommended in vivo formulation examples.
  • Store protected from light, under inert gas; stability varies by storage temperature.

Catalog No. 50-002-03466 Supplier Medchemexpress LLC Supplier No. HY12650710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-amido-PEG1-C2-NHS ester is a noncleavable ADC linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester, enabling conjugation to thiols and primary amines for antibody-drug conjugate construction and bioconjugation.

  • Contains maleimide and NHS ester functional groups for thiol and amine coupling.
  • Molecular weight approximately 381.34 g/mol.
  • Chemical formula C16H19N3O8; white to off-white solid.
  • High purity (listed as 99.69%) suitable for research applications.
  • Soluble in DMSO (100 mg/mL) with recommended in vivo formulation examples.
  • Store protected from light, under inert gas; stability varies by storage temperature.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.