Learn More
Medchemexpress LLC Mal-amido-PEG1-C2-NHS ester | 1260092-50-9 | 99.7% | 381.34 g/mol | C16H19N3O8 | 25 MG
SDP
Catalog No. 5000202447
Encompass_Preferred
Mal-amido-PEG1-C2-NHS ester is a heterobifunctional PEG1-based linker containing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is designed for bioconjugation applications, enabling selective labeling of amines and thiols and supplied as a solid with recommended solubility in DMSO.
  • Heterobifunctional maleimide-NHS linker for bioconjugation.
  • Enables amine labeling via NHS ester and thiol conjugation via maleimide.
  • High purity suitable for research applications and ADC linker chemistry.
  • Available in small pack sizes for laboratory use and soluble in DMSO.
  • Molecular weight 381.34 g/mol and CAS 1260092-50-9.
Catalog No. 50-002-02447 Supplier Medchemexpress LLC Supplier No. HY12650725MG
Mal-amido-PEG1-C2-NHS ester is a heterobifunctional PEG1-based linker containing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is designed for bioconjugation applications, enabling selective labeling of amines and thiols and supplied as a solid with recommended solubility in DMSO.
  • Heterobifunctional maleimide-NHS linker for bioconjugation.
  • Enables amine labeling via NHS ester and thiol conjugation via maleimide.
  • High purity suitable for research applications and ADC linker chemistry.
  • Available in small pack sizes for laboratory use and soluble in DMSO.
  • Molecular weight 381.34 g/mol and CAS 1260092-50-9.