Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-PEG10-C2-NHS ester | 1137109-22-8 | 98.0% | 777.81 | C34H55N3O17 | 1 G
SDP

Catalog No. 5000202448
Encompass_Preferred

Mal-amido-PEG10-C2-NHS ester is a non-cleavable ADC linker containing a maleimide group for thiol conjugation and an N-hydroxysuccinimide (NHS) ester for labeling primary amines. It is used in bioconjugation workflows such as antibody-drug conjugate assembly and protein or oligonucleotide labeling.

  • Contains maleimide and NHS ester functional groups.
  • Enables thiol- and amine-targeted conjugation chemistries.
  • Supplied as a solid in multiple package sizes including 1 g.
  • Molecular weight of 777.81 and formula C34H55N3O17.
  • Provided for research use in biochemical and ADC development applications.

Catalog No. 50-002-02448 Supplier Medchemexpress LLC Supplier No. HY1265091G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-amido-PEG10-C2-NHS ester is a non-cleavable ADC linker containing a maleimide group for thiol conjugation and an N-hydroxysuccinimide (NHS) ester for labeling primary amines. It is used in bioconjugation workflows such as antibody-drug conjugate assembly and protein or oligonucleotide labeling.

  • Contains maleimide and NHS ester functional groups.
  • Enables thiol- and amine-targeted conjugation chemistries.
  • Supplied as a solid in multiple package sizes including 1 g.
  • Molecular weight of 777.81 and formula C34H55N3O17.
  • Provided for research use in biochemical and ADC development applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.