Learn More
Medchemexpress LLC Mal-amido-PEG10-C2-NHS ester | 1137109-22-8 | MFCD20226950 | 777.81 g/mol | C34H55N3O17 | 250 MG
SDP
Catalog No. 5000201892
Encompass_Preferred
Mal-amido-PEG10-C2-NHS ester is a PEG-based, non-cleavable linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. The NHS ester enables conjugation to primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules, while the maleimide allows thiol-selective coupling. This reagent is supplied for research use only.
  • Non-cleavable PEG-based linker with maleimide and NHS ester functionality.
  • Enables amine and thiol bioconjugation for protein and oligonucleotide labeling.
  • Molecular weight approximately 777.8 g/mol for predictable mass additions.
  • Provided as a small-molecule reagent suitable for research applications.
  • Store in a tightly closed container in a cool, dry, well-ventilated place.
Catalog No. 50-002-01892 Supplier Medchemexpress LLC Supplier No. HY126509250MG
Mal-amido-PEG10-C2-NHS ester is a PEG-based, non-cleavable linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. The NHS ester enables conjugation to primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules, while the maleimide allows thiol-selective coupling. This reagent is supplied for research use only.
  • Non-cleavable PEG-based linker with maleimide and NHS ester functionality.
  • Enables amine and thiol bioconjugation for protein and oligonucleotide labeling.
  • Molecular weight approximately 777.8 g/mol for predictable mass additions.
  • Provided as a small-molecule reagent suitable for research applications.
  • Store in a tightly closed container in a cool, dry, well-ventilated place.