Learn More
Medchemexpress LLC Mal-amido-PEG12-acid | 2378428-27-2 | C34H60N2O17 | 50 MG
SDP
Catalog No. 5000355409
Encompass_Preferred
Mal-amido-PEG12-acid is a PEG-based PROTAC linker suitable for synthesizing PROTACs. It functions by connecting two distinct ligands, one targeting an E3 ubiquitin ligase and the other for the specific target protein, allowing for the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • Can be used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-003-55409 Supplier Medchemexpress LLC Supplier No. HY14097450MG
Mal-amido-PEG12-acid is a PEG-based PROTAC linker suitable for synthesizing PROTACs. It functions by connecting two distinct ligands, one targeting an E3 ubiquitin ligase and the other for the specific target protein, allowing for the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • Can be used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.